Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-6-chloropyridazine-4-carboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

073975-1G 073975-5G
102533-990EA 187.69 USD InStock
102533-990 102533-992
Methyl-6-chloropyridazine-4-carboxylate ≥95%
Methyl-6-chloropyridazine-4-carboxylate
Hazard: Irritant


Formula: C₆H₅ClN₂O₂
MW: 172.5691 g/mol
MDL Number: MFCD16660968
CAS Number: 1093860-48-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty