Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-p-toluic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

074147-100G 074147-25G
102534-896EA 118.46 USD InStock
102534-896 102663-980
3-Amino-p-toluic acid ≥95%
3-Amino-p-toluic acid
Hazard: Irritant


Formula: C₈H₉NO₂
MDL Number: MFCD00007742
CAS Number: 2458-12-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty