Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4,6-dimethoxypyrimidine ≥95%

Supplier: MATRIX SCIENTIFIC MS

074209-10G 074209-100G
102535-220EA 72.31 USD InStock
102535-220 102535-222
2-Amino-4,6-dimethoxypyrimidine ≥95%
2-Amino-4,6-dimethoxypyrimidine
Hazard: Irritant


Formula: C₆H₉N₃O₂
MDL Number: MFCD00038832
CAS Number: 36315-01-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty