Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-fluoro-5-nitrobenzoic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

074686-1G 074686-5G 074686-10G
102537-694EA 213.85 USD InStock
102537-694 102537-696 102537-698
2-Chloro-4-fluoro-5-nitrobenzoic acid ≥95%
2-Chloro-4-fluoro-5-nitrobenzoic acid
Hazard: Irritant


Formula: C₇H₃ClFNO
MDL Number: MFCD04115715
CAS Number: 114776-15-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty