Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(-)-β-Aminophenethyl alcohol ≥95%

Supplier: MATRIX SCIENTIFIC MS

074840-100G 074840-25G
102538-492EA 306.15 USD InStock
102538-492 102833-724
(R)-(-)-β-Aminophenethyl alcohol ≥95%
(R)-(-)-β-Aminophenethyl alcohol
Hazard: Irritant/Stored under Argon/Store in Freezer


Formula: C₈H₁₁NO
Melting Pt: 76…78 °C
MDL Number: MFCD00008062
CAS Number: 56613-80-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty