Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Iodoxybenzoic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

074939-100G 074939-25G
102538-956EA 273.85 USD InStock
102538-956 102833-746
2-Iodoxybenzoic acid ≥95%
2-Iodoxybenzoic acid
Hazard: Corrosive


Formula: C₇H₅IO₄
Melting Pt: 250 °C (dec.)
MDL Number: MFCD02912492
CAS Number: 61717-82-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty