Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4'-Disulfanediylbis(2-aminobutanoic acid) ≥95%

Supplier: MATRIX SCIENTIFIC MS

074962-5G 074962-10G 074962-100G
102539-068EA 53.85 USD InStock
102539-068 102539-070 102539-072
4,4'-Disulfanediylbis(2-aminobutanoic acid) ≥95%
4,4'-Disulfanediylbis(2-aminobutanoic acid)
Hazard: Irritant


Formula: C₈H₁₆N₂O₄S₂
MDL Number: MFCD00063095
CAS Number: 462-10-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty