Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Pyridone ≥95%

Supplier: MATRIX SCIENTIFIC MS

074987-10G 074987-100G
102539-192EA 15.38 USD InStock
102539-192 102539-194
3-Pyridone ≥95%
3-Pyridone
Hazard: Irritant


Formula: C₅H₅NO
Melting Pt: 125-128°C
MDL Number: MFCD00006378
CAS Number: 109-00-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty