Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxy-4-iodobenzoic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

074998-1G 074998-5G 074998-10G
102539-246EA 284.62 USD InStock
102539-246 102539-248 102539-250
3-Hydroxy-4-iodobenzoic acid ≥95%
3-Hydroxy-4-iodobenzoic acid
Hazard: Irritant


Formula: C₇H₅IO₃
MW: 264.02
MDL Number: MFCD02068386
CAS Number: 58123-77-6
UN: 2811
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty