Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-m-anisidine ≥95%

Supplier: MATRIX SCIENTIFIC MS

075191-5G 075191-25G
102540-228EA 52.31 USD InStock
102540-228 102833-792
4-Chloro-m-anisidine ≥95%
4-Chloro-m-anisidine
Hazard: Toxic


Formula: C₇H₈ClNO
MW: 157.6
Melting Pt: 78…81 °C
MDL Number: MFCD00672967
CAS Number: 13726-14-2
UN: 2233
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty