Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Aminopiperidine-2,6-dione hydrochloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

075215-1G 075215-5G 075215-25G
102540-336EA 29.23 USD InStock
102540-336 102540-338 102551-946
3-Aminopiperidine-2,6-dione hydrochloride ≥95%
3-Aminopiperidine-2,6-dione hydrochloride
Hazard: Irritant/Hygroscopic


Formula: C5H9ClN2O2
MW: 164.59 g/mol
Melting Pt: 245 °C
MDL Number: MFCD11042437
CAS Number: 24666-56-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty