Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzofuran ≥95%

Supplier: MATRIX SCIENTIFIC MS

075261-100G 075261-25G
102540-550EA 127.69 USD InStock
102540-550 102833-796
Benzofuran ≥95%
Benzofuran
Hazard: Flammable


Formula: C₈H₆O
Boiling Pt: 173…175 °C
Density: 1.095
Flash Pt: 56 °C
MDL Number: MFCD00005847
CAS Number: 271-89-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty