Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Aminoethoxy)benzene ≥95%

Supplier: MATRIX SCIENTIFIC MS

075275-5G 075275-25G
102540-616EA 84.62 USD InStock
102540-616 102540-618
(2-Aminoethoxy)benzene ≥95%
(2-Aminoethoxy)benzene
Hazard: Corrosive/Air Sensitive


Formula: C₆H₅OCH₂CH₂NH₂
Boiling Pt: 88-90°C/1mm
Density: 1.048
Flash Pt: 107°C
MDL Number: MFCD00052975
CAS Number: 1758-46-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty