Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-methoxypyridine hydrochloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

075297-250MG 075297-1G 075297-5G
102540-722EA 456.92 USD InStock
102540-722 102540-724 102540-726
4-Bromo-3-methoxypyridine hydrochloride ≥95%
4-Bromo-3-methoxypyridine hydrochloride
Hazard: Irritant


Formula: C₆H₇BrClNO
MDL Number: MFCD11226828
CAS Number: 1209335-53-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty