Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-2-Methyltetrahydrofuran-3-one ≥95%

Supplier: MATRIX SCIENTIFIC MS

075308-10G 075308-100G
102540-774EA 58.46 USD InStock
102540-774 102540-776
(±)-2-Methyltetrahydrofuran-3-one ≥95%
(±)-2-Methyltetrahydrofuran-3-one
Hazard: Irritant


Formula: C₅H₈O₂
MDL Number: MFCD00010423
CAS Number: 3188-00-9
UN: 1224
ADR: 3,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty