Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trityl candesartan ≥95%

Supplier: MATRIX SCIENTIFIC MS

075316-250MG 075316-1G
102540-810EA 400 USD InStock
102540-810 102540-812
Trityl candesartan ≥95%
Trityl candesartan
Hazard: Irritant


Formula: C₄₃H₃₄N₆O₃
MW: 682.7685 g/mol
MDL Number: MFCD08436154
CAS Number: 139481-72-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty