Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dichlorobenzyl alcohol ≥95%

Supplier: MATRIX SCIENTIFIC MS

075347-10G 075347-100G
102540-940EA 107.69 USD InStock
102540-940 102540-942
3,4-Dichlorobenzyl alcohol ≥95%
3,4-Dichlorobenzyl alcohol
Hazard: Irritant


Formula: C₇H₆Cl₂O
MW: 177.03 g/mol
Boiling Pt: 271
MDL Number: MFCD00004633
CAS Number: 1805-32-9
UN: 2811
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty