Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Salicylonitrile ≥95%

Supplier: MATRIX SCIENTIFIC MS

075375-10G 075375-100G 075375-500G
102541-066EA 23.08 USD InStock
102541-066 102541-068 102541-070
Salicylonitrile ≥95%
Salicylonitrile
Hazard: Irritant


Formula: C₇H₅NO
MW: 119.12 g/mol
Boiling Pt: 273
Melting Pt: 92…95 °C
MDL Number: MFCD00002145
CAS Number: 611-20-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty