Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2,4-Dichlorophenyl)acetic acid methyl ester ≥95%

Supplier: MATRIX SCIENTIFIC MS

075402-10G 075402-100G
102541-190EA 143.08 USD InStock
102541-190 102541-192
(2,4-Dichlorophenyl)acetic acid methyl ester ≥95%
(2,4-Dichlorophenyl)acetic acid methyl ester
Hazard: Irritant


Formula: (C₆H₃Cl₂)CH₂COOCH₃
MDL Number: MFCD00018381
CAS Number: 55954-23-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty