Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxybenzyl alcohol ≥95%

Supplier: MATRIX SCIENTIFIC MS

075405-100G 075405-500G
102541-204EA 178.46 USD InStock
102541-204 102541-206
4-Hydroxybenzyl alcohol ≥95%
4-Hydroxybenzyl alcohol
Hazard: Irritant


Formula: C₇H₈O₂
MDL Number: MFCD00004658
CAS Number: 623-05-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty