Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Nitrobenzyl alcohol ≥95%

Supplier: MATRIX SCIENTIFIC MS

075421-100G 075421-500G
102541-282EA 67.69 USD InStock
102541-282 102664-044
2-Nitrobenzyl alcohol ≥95%
2-Nitrobenzyl alcohol
Hazard: Irritant-Harmful


Formula: C₇H₇NO₃
Melting Pt: 69…74 °C
MDL Number: MFCD00007186
CAS Number: 612-25-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty