Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

p-Tolylhydrazinium chloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

075437-25G 075437-100G
102541-350EA 26.15 USD InStock
102541-350 102541-352
p-Tolylhydrazinium chloride ≥95%
p-Tolylhydrazinium chloride
Hazard: Toxic


Formula: C₇H₁₁ClN₂
Melting Pt: ∼ 240 °C (dec.)
MDL Number: MFCD00012940
CAS Number: 637-60-5
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty