Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Bromophenyl)acetonitrile ≥95%

Supplier: MATRIX SCIENTIFIC MS

075473-10G 075473-100G
102541-516EA 91.54 USD InStock
102541-516 102541-518
(3-Bromophenyl)acetonitrile ≥95%
(3-Bromophenyl)acetonitrile
Hazard: Toxic


Formula: C₈H₆BrN
Boiling Pt: 275 °C
Melting Pt: 25…28 °C
Flash Pt: 126 °C
MDL Number: MFCD00001906
CAS Number: 31938-07-5
UN: 3449
ADR: 6.1,I
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty