Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diphenyl sulphoxide ≥95%

Supplier: MATRIX SCIENTIFIC MS

075533-10G 075533-100G
102541-786EA 72.31 USD InStock
102541-786 102541-788
Diphenyl sulphoxide ≥95%
Diphenyl sulphoxide
Hazard: Irritant


Formula: C₁₂H₁₀OS
MDL Number: MFCD00002085
CAS Number: 945-51-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty