Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzothiazol-2-ol ≥95%

Supplier: MATRIX SCIENTIFIC MS

075759-5G 075759-10G 075759-100G
102542-888EA 107.69 USD InStock
102542-888 102542-890 102542-892
Benzothiazol-2-ol ≥95%
Benzothiazol-2-ol
Hazard: Irritant


Formula: C₇H₅NOS
MDL Number: MFCD00022868
CAS Number: 934-34-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty