Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chlorophenyl methyl sulphone ≥95%

Supplier: MATRIX SCIENTIFIC MS

075808-5G 075808-10G 075808-100G
102543-102EA 107.69 USD InStock
102543-102 102543-104 102543-106
4-Chlorophenyl methyl sulphone ≥95%
4-Chlorophenyl methyl sulphone
Hazard: Irritant


Formula: C₇H₇ClO₂S
MW: 190.65 g/mol
MDL Number: MFCD00025067
CAS Number: 98-57-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty