Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(-)-3-Chloro-1,2-propanediol ≥95%

Supplier: MATRIX SCIENTIFIC MS

075856-100G 075856-25G
102543-328EA 169.23 USD InStock
102543-328 102833-880
(R)-(-)-3-Chloro-1,2-propanediol ≥95%
(R)-(-)-3-Chloro-1,2-propanediol
Hazard: Toxic/Store in Freezer


Formula: ClCH₂CHOHCH₂OH
Boiling Pt: 213°C
Density: 1.322
Flash Pt: 137°C
MDL Number: MFCD00135169
CAS Number: 57090-45-6
UN: 2689
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty