Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lomefloxacin hydrochloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

075878-5G 075878-10G 075878-100G
102543-428EA 90.77 USD InStock
102543-428 102543-430 102543-432
Lomefloxacin hydrochloride ≥95%
Lomefloxacin hydrochloride
Hazard: Irritant


Formula: C₁₇H₁₉F₂N₃O₃·HCl
MW: 387.82 g/mol
MDL Number: MFCD00214312
CAS Number: 98079-52-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty