Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxypyridine ≥95%

Supplier: MATRIX SCIENTIFIC MS

075906-100G 075906-25G
102543-570EA 183.08 USD InStock
102543-570 102664-074
4-Methoxypyridine ≥95%
4-Methoxypyridine
Hazard: Irritant


Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 191 °C
Melting Pt: 4 °C
Density: 1.075
Flash Pt: 75 °C
MDL Number: MFCD00674049
CAS Number: 620-08-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty