Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-((2'-(1H-Tetrazol-5-yl)[1,1'-biphenyl]-4-yl)methyl)-2-butyl-1,3-diazaspiro[4.4]non-1-en-4-one ≥95%

Supplier: MATRIX SCIENTIFIC MS

075934-1G 075934-5G 075934-10G
102543-704EA 30.77 USD InStock
102543-704 102543-706 102543-708
3-((2'-(1H-Tetrazol-5-yl)[1,1'-biphenyl]-4-yl)methyl)-2-butyl-1,3-diazaspiro[4.4]non-1-en-4-one ≥95%
3-((2'-(1H-Tetrazol-5-yl)[1,1'-biphenyl]-4-yl)methyl)-2-butyl-1,3-diazaspiro[4.4]non-1-en-4-one
Hazard: Irritant


Formula: C₂₅H₂₈N₆O
MDL Number: MFCD00864464
CAS Number: 138402-11-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty