Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-4-chloronicotinate ≥95%

Supplier: MATRIX SCIENTIFIC MS

075953-1G 075953-5G
102543-786EA 523.08 USD InStock
102543-786 102543-788
Ethyl-4-chloronicotinate ≥95%
Ethyl-4-chloronicotinate
Hazard: Irritant/Light Sensitive/Stored under Argon


Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
Boiling Pt: 85 °C (1 mmHg)
MDL Number: MFCD05664028
CAS Number: 37831-62-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty