Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-(Bromomethyl)-1H-indazole hydrobromide ≥95%

Supplier: MATRIX SCIENTIFIC MS

075978-250MG 075978-1G
102543-920EA 598.46 USD InStock
102543-920 102543-922
6-(Bromomethyl)-1H-indazole hydrobromide ≥95%
6-(Bromomethyl)-1H-indazole hydrobromide
Hazard: Irritant


Formula: C₈H₈Br₂N₂
MDL Number: MFCD14586465
CAS Number: 368426-63-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty