Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N1,N1,N2-Trimethylethane-1,2-diamine dihydrochloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

075983-1G 075983-5G
102543-944EA 569.23 USD InStock
102543-944 102543-946
N1,N1,N2-Trimethylethane-1,2-diamine dihydrochloride ≥95%
N1,N1,N2-Trimethylethane-1,2-diamine dihydrochloride
Hazard: Irritant


Formula: C₅H₁₆Cl₂N₂
MDL Number: MFCD21604009
CAS Number: 326888-32-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty