Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-6-chloropurine riboside ≥95%

Supplier: MATRIX SCIENTIFIC MS

076001-1G 076001-5G 076001-10G
102544-038EA 112.31 USD InStock
102544-038 102544-040 102544-042
2-Amino-6-chloropurine riboside ≥95%
2-Amino-6-chloropurine riboside
Hazard: Irritant


Formula: C₁₀H₁₂ClN₅O₄
MW: 301.69 g/mol
Melting Pt: 171…172 °C (dec.)
MDL Number: MFCD00005735
CAS Number: 2004-07-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty