Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-fluorobiphenyl ≥95%

Supplier: MATRIX SCIENTIFIC MS

076116-10G 076116-100G
102544-568EA 55.38 USD InStock
102544-568 102544-570
4-Bromo-2-fluorobiphenyl ≥95%
4-Bromo-2-fluorobiphenyl
Hazard: Irritant


Formula: C₁₂H₈BrF
MDL Number: MFCD00051716
CAS Number: 41604-19-7
UN: 3152
ADR: 9,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty