Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Norbornene-2-carboxylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

076119-10G 076119-100G
102544-582EA 72.31 USD InStock
102544-582 102544-584
5-Norbornene-2-carboxylic acid ≥95%
5-Norbornene-2-carboxylic acid
Hazard: Corrosive


Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 136…138 °C (14 mmHg)
Density: 1.129
MDL Number: MFCD00085356
CAS Number: 120-74-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty