Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Isopropylaniline ≥95%

Supplier: MATRIX SCIENTIFIC MS

076138-10G 076138-100G
102544-662EA 43.08 USD InStock
102544-662 102544-664
N-Isopropylaniline ≥95%
N-Isopropylaniline
Hazard: Irritant


Formula: C₆H₅NHCH(CH₃)₂
MDL Number: MFCD00026347
CAS Number: 768-52-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25a6ef05d2a86c1f6dda0fa1fd119d04c095270c&vid=25a6ef05d2a86c1f6dda0fa1fd119d04c095270c&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty