Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-4-chlorobenzoic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

076156-10G 076156-100G
102544-742EA 15.38 USD InStock
102544-742 102544-744
3-Amino-4-chlorobenzoic acid ≥95%
3-Amino-4-chlorobenzoic acid
Hazard: Irritant


Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 330
Melting Pt: 213…218 °C
MDL Number: MFCD00007671
CAS Number: 2840-28-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty