Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methylguanidine hydrochloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

076180-5G 076180-10G 076180-100G
102544-860EA 86.15 USD InStock
102544-860 102544-862 102544-864
1-Methylguanidine hydrochloride ≥95%
1-Methylguanidine hydrochloride
Hazard: Irritant


Formula: C₂H₈ClN₃
MDL Number: MFCD00012576
CAS Number: 21770-81-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty