Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3,4-dihydroxybenzoate ≥95%

Supplier: MATRIX SCIENTIFIC MS

076188-25G 076188-100G
102833-908EA 176.92 USD InStock
102833-908 102833-910
Methyl-3,4-dihydroxybenzoate ≥95%
Methyl-3,4-dihydroxybenzoate
Hazard: Irritant


Formula: C₈H₈O₄
MW: 168.15 g/mol
Melting Pt: 134…138 °C
MDL Number: MFCD00016437
CAS Number: 2150-43-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty