Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl isobutyrylacetate ≥95%

Supplier: MATRIX SCIENTIFIC MS

076193-100G 076193-25G
102544-926EA 444.62 USD InStock
102544-926 102833-914
Ethyl isobutyrylacetate ≥95%
Ethyl isobutyrylacetate
Hazard: Flammable


Formula: C₈H₁₄O₃
MW: 158.2 g/mol
Boiling Pt: 172…174 °C
Density: 0.98
Flash Pt: 53 °C
MDL Number: MFCD00009198
CAS Number: 7152-15-0
UN: 3272
ADR: 3 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty