Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydrazinylbenzonitrile hydrochloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

076289-5G 076289-25G
102545-414EA 158.46 USD InStock
102545-414 102664-092
3-Hydrazinylbenzonitrile hydrochloride ≥95%
3-Hydrazinylbenzonitrile hydrochloride


Formula: C7H8ClN3
MDL Number: MFCD03094642
CAS Number: 2881-99-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty