Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Aminocyclopentanol hydrochloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

076323-250MG 076323-1G
102545-578EA 640 USD InStock
102545-578 102545-580
3-Aminocyclopentanol hydrochloride ≥95%
3-Aminocyclopentanol hydrochloride


Formula: C₅H₁₂ClNO
MDL Number: MFCD16619116
CAS Number: 1184919-69-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty