Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-5-oxazolecarboxylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

076342-250MG 076342-1G 076342-5G
102545-668EA 2544.62 USD InStock
102545-668 102545-670 102545-672
2-Bromo-5-oxazolecarboxylic acid ≥95%
2-Bromo-5-oxazolecarboxylic acid
Hazard: Irritant


Formula: C₄H₂BrNO₃
MDL Number: MFCD21604290
CAS Number: 1373253-25-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty