Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Propyzamide ≥95%

Supplier: MATRIX SCIENTIFIC MS

076346-1G 076346-5G 076346-10G
102545-688EA 90.77 USD InStock
102545-688 102545-690 102545-692
Propyzamide ≥95%
Propyzamide
Hazard: Irritant


Formula: C₁₂H₁₁Cl₂NO
MDL Number: MFCD00055346
CAS Number: 23950-58-5
UN: 3077
ADR: 9,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty