Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N2,N2-Dimethylpyridine-2,5-diamine dihydrochloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

076416-250MG 076416-1G 076416-5G
102546-072EA 640 USD InStock
102546-072 102546-074 102546-076
N2,N2-Dimethylpyridine-2,5-diamine dihydrochloride ≥95%
N2,N2-Dimethylpyridine-2,5-diamine dihydrochloride
Hazard: Irritant


Formula: C₇H₁₃Cl₂N₃
MDL Number: MFCD09908196
CAS Number: 26878-31-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty