Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Imidazo[2,1-b]thiazole-5-carboxylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

076463-250MG 076463-1G
102546-320EA 1053.85 USD InStock
102546-320 102546-322
Imidazo[2,1-b]thiazole-5-carboxylic acid ≥95%
Imidazo[2,1-b]thiazole-5-carboxylic acid
Hazard: Irritant


Formula: C₆H₄N₂O₂S
MW: 168.17 g/mol
MDL Number: MFCD09743157
CAS Number: 17782-81-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty