Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2H-1,2,3-Triazole-2-acetic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

076486-250MG 076486-1G
102546-448EA 355.38 USD InStock
102546-448 102546-450
2H-1,2,3-Triazole-2-acetic acid ≥95%
2H-1,2,3-Triazole-2-acetic acid
Hazard: Irritant


Formula: C₄H₅N₃O₂
MDL Number: MFCD03840810
CAS Number: 4320-91-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty