Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Thiazol-2-yl)aniline ≥95%

Supplier: MATRIX SCIENTIFIC MS

076511-250MG 076511-1G 076511-5G
102546-586EA 355.38 USD InStock
102546-586 102546-588 102546-590
4-(Thiazol-2-yl)aniline ≥95%
4-(Thiazol-2-yl)aniline
Hazard: Irritant


Formula: C₉H₈N₂S
MW: 176.24 g/mol
MDL Number: MFCD06655097
CAS Number: 193017-26-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty