Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methoxynicotinimidamide hydrochloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

076591-250MG 076591-1G 076591-5G
102546-992EA 355.38 USD InStock
102546-992 102546-994 102546-996
6-Methoxynicotinimidamide hydrochloride ≥95%
6-Methoxynicotinimidamide hydrochloride


Formula: C₇H₁₀ClN₃O
MDL Number: MFCD08234869
CAS Number: 201937-22-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty